missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ TACS TdT In Situ Apoptosis Detection Kit, Blue Label

A TUNEL assay designed for fixed cells, embedded tissue, and frozen tissue using light and electron microscopy

Brand:  R&D Systems™ 4810-30-K

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13445104

  • £440.72 / 30 tests

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Includes: Proteinase K Solution; Cytonin™; TdT enzyme, dNTP mix and buffers; Strep-HRP; TACS-Blue Label; Blue Streptavidin-Diluent; Cations; TACS-Nuclease™ and buffer; Nuclear Fast Red

Description

Description

Contains a highly purified form of the TdT enzyme for the enzymatic incorporation of biotinylated nucleotides
Biotin labeling is achieved using streptavidin-horseradish peroxidase and the colorimetric substrate TACS Blue Label

Specifications

Specifications

30 Tests
DAB
The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
TACS TdT In Situ Apoptosis Detection Kit
TUNEL for fixed cells, embedded tissue, frozen tissue using light and electron microscopy; Reagents to detect DNA fragmentn in situ
Videos