missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rhesus Macaque TNF-alpha Recombinant Protein

Brand:  R&D Systems™ 1070-RM-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458443

  • £240.25 / 25µg


Estimated Shipment
27-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Val77-Leu233
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
TNF-alpha
25μg
<0.01 EU per 1μg of the protein by the LAL method.
Yes
17.4 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rhesus Macaque
Unlabeled
Videos