missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rhesus Macaque IL-4 Recombinant Protein

Brand:  R&D Systems™ 1577-IL-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459703

  • £273.25 / 10µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, His25-Ser153, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
IL-4
10μg
Unlabeled
15.1 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rhesus Macaque
Yes
Videos