missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rhesus Macaque IL-18/IL-1F4 Recombinant Protein

Brand:  R&D Systems™ 2548-RM-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461444

  • £265.33 / 25µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Tyr37-Asp193
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.2 kDa
Formulation: Supplied as a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT.
Regulatory Status: RUO

Specifications

Specifications

Reconstitute at 25μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
18.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rhesus Macaque
Yes
Lyophilized from a 0.2μm filtered solution in Tris, NaCl, EDTA, TCEP and PEG 8000 with BSA as a carrier protein.
IL-18/IL-1F4
25μg
Unlabeled
Videos