missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Rhesus Macaque IL-18/IL-1F4 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Conjugate | Unconjugated |
---|---|
Molecular Weight (g/mol) | 18.2 kDa |
Name | IL-18/IL-1F4 |
Quantity | 25μg |
Recombinant | Yes |
Description
The Recombinant Rhesus Macaque IL-18/IL-1F4 Protein is derived from E. coli. The Recombinant Rhesus Macaque IL-18/IL-1F4 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
IL-18/IL-1F4 | |
Yes |
18.2 kDa | |
25μg | |
E. coli-derived rhesus macaque IL-18/IL-1F4 protein Tyr37-Asp193 |