Learn More
R&D Systems™ Rhesus Macaque IL-12 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 3216-RL-025
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13432584
  • £458.64 / 25µg
Estimated Shipment date
from Supplier 10-03-2020
Description
Description
The Recombinant Rhesus Macaque IL-12 Protein is derived from CHO. The Recombinant Rhesus Macaque IL-12 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
IL-12
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
35.6 kDa (p40), 23.4 kDa (p35)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rhesus Macaque
Disulfide-linked heterodimer
Yes