missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat TNF-alpha Recombinant Protein

Brand:  R&D Systems™ 510-RT-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465294

  • £490.12 / 50µg


Estimated Shipment
03-09-2018
Add to basket

Description

Description

Source: E. coli-derived, Leu80-Leu235, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and DTT.
Entrez Gene ID: 24835
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS and DTT.
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Unlabeled
24835
TNF-alpha
50μg
<0.1 EU per 1μg of the protein by the LAL method.
Yes
Videos