missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat Pentraxin 2/SAP Recombinant Protein

Brand:  R&D Systems™ 1895-SA-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460194

  • £316.81 / 50µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Gln21-Ser228 with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 25 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, NaCl and CaCl2.
Entrez Gene ID: 29339
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in MOPS, NaCl and CaCl2.
25 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Unlabeled
29339
Pentraxin 2/SAP
50μg
Noncovalently-linked homopentamer
Yes
Videos