missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat Pentraxin 2/SAP Recombinant Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 1895-SA-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13450194

  • £352.80 / 50µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Rat Pentraxin 2/SAP Protein is derived from NS0. The Recombinant Rat Pentraxin 2/SAP Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 with BSA as a carrier protein.
25 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Noncovalently-linked homopentamer
Yes
29339
Pentraxin 2/SAP
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents