missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat PDGF-BB Recombinant Protein

Brand:  R&D Systems™ 520-BB-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465344

  • £289.09 / 50µg


Estimated Shipment
02-01-2019
Add to basket

Description

Description

Source: E. coli-derived, Ser74-Thr182
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 25 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 24628
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
25 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Unlabeled
24628
PDGF-BB Homodimer
50μg
Disulfide-linked homodimer
Yes
Videos