missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat IL-9 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Molecular Weight (g/mol) | 14.1 kDa |
---|---|
Name | IL-9 |
Quantity | 10μg |
Source | Mouse myeloma cell line,NS0-derived rat IL-9 protein Gln19-Ala144 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Description
The Recombinant Rat IL-9 Protein is derived from NS0. The Recombinant Rat IL-9 Protein has been validated for the following applications: Bioactivity.Specifications
14.1 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
IL-9 | |
Mouse myeloma cell line,NS0-derived rat IL-9 protein Gln19-Ala144 | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |