missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Rat IL-5 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

POA

Specifications

Gene ID (Entrez) 24497
Molecular Weight (g/mol) 13 kDa (monomer)
Name IL-5
Quantity 10μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived rat IL-5 protein Met20-Val132
View More Specs

Description

Description

Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.05-0.2 ng/mL.
Specifications

Specifications

24497
IL-5
Spodoptera frugiperda,Sf 21 (baculovirus)-derived rat IL-5 protein Met20-Val132
Disulfide-linked homodimer
Recombinant
13 kDa (monomer)
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Videos
SDS
Documents

Documents

Product Certifications