missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IL-4 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 504-RL-005

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13445264

  • £256.04 / 5µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Rat IL-4 Protein is derived from E. coli. The Recombinant Rat IL-4 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
13.5 kDa
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived rat IL-4 protein Cys25-Ser147,with an N-terminal Met
B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Yes
287287
IL-4
5μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Videos
Documents