missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IL-4 Recombinant Protein

Brand:  R&D Systems™ 504-RL-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465264

  • £631.00 / 25µg


Estimated Shipment
29-06-2018
Add to basket

Description

Description

Source: E. coli-derived, Cys25-Ser147, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 287287
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
13.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Yes
287287
IL-4
25μg
Unlabeled
Videos