missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IL-3 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 2524-RL-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13441394

  • £284.16 / 25µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Rat IL-3 Protein is derived from E. coli. The Recombinant Rat IL-3 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
16.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived rat IL-3 protein Ile27-Cys169,with an N-terminal Met
Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor
Yes
24495
IL-3
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Videos
Documents