missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Rat IL-3 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 24495 |
Molecular Weight (g/mol) | 16.3 kDa |
Name | IL-3 |
Quantity | 25μg |
Description
The Recombinant Rat IL-3 Protein is derived from E. coli. The Recombinant Rat IL-3 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
16.3 kDa | |
25μg | |
E. coli-derived rat IL-3 protein Ile27-Cys169,with an N-terminal Met |
24495 | |
IL-3 | |
Yes |