missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IL-23 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 3136-RL-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13432504

  • £352.80 / 10µg

Estimated Shipment date
from Supplier 02-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Rat IL-23 Protein is derived from Sf 21 (baculovirus). The Recombinant Rat IL-23 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl.
IL-23
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
58.3 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
<0.01 EU per 1μg of the protein by the LAL method.
Yes
Videos
Documents