missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat IL-23 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Molecular Weight (g/mol) | 58.3 kDa |
---|---|
Name | IL-23 |
Quantity | 10μg |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived rat IL-23 protein, (N-terminus) Rat IL-23 p40 (Met23-Ser335) Accession &Num; NP_072133 IGSGSSRGGSGSGGSGGGGSK Rat IL-23 p19 (Leu20-Ala196) Accession &Num; NP_569094 8-His tag (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Description
The Recombinant Rat IL-23 Protein is derived from Sf 21 (baculovirus). The Recombinant Rat IL-23 Protein has been validated for the following applications: Bioactivity.Specifications
58.3 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
IL-23 | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived rat IL-23 protein, (N-terminus) Rat IL-23 p40 (Met23-Ser335) Accession &Num; NP_072133 IGSGSSRGGSGSGGSGGGGSK Rat IL-23 p19 (Leu20-Ala196) Accession &Num; NP_569094 8-His tag (C-terminus) | |
<0.01 EU per 1μg of the protein by the LAL method. | |
Recombinant |