missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IL-22 Recombinant Protein

Brand:  R&D Systems™ 1316-RL-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459123

  • £364.33 / 10µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Leu27-Val172, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 500836
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
16.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Unlabeled
500836
IL-22
10μg
<0.1 EU per 1μg of the protein by the LAL method.
Yes
Videos