missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IL-12 Recombinant Protein

Brand:  R&D Systems™ 1760-RL-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459943

  • £323.41 / 10ยตg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Met23-Ser335 (p40) & Arg23-Ser215 (p35)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36.8 kDa (p40), 22.6 kDa (p35)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 84405
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
36.8 kDa (p40), 22.6 kDa (p35)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
<0.1 EU per 1μg of the protein by the LAL method.
Yes
84405
IL-12
10μg
Disulfide-linked heterodimer
Unlabeled
Videos