missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IL-12 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1760-RL-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13469943

  • £370.44 / 10µg

Estimated Shipment date
from Supplier 03-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Rat IL-12 Protein is derived from Sf 21 (baculovirus). The Recombinant Rat IL-12 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
36.8 kDa (p40), 22.6 kDa (p35)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Disulfide-linked heterodimer
Unlabeled
84405
IL-12
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
<0.1 EU per 1μg of the protein by the LAL method.
Yes
Videos
Documents