missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IL-10 Recombinant Protein

Brand:  R&D Systems™ 522-RL-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13445354

  • £767.96 / 25µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Source: E. coli-derived, Ser19-Asn178
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19 kDa
Formulation: Supplied as a 0.2μm filtered solution in NaH2PO4 and NaCl.
Entrez Gene ID: 25325
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein.
19 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Yes
25325
IL-10
25μg
Unlabeled
Videos