missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IL-1 beta/IL-1F2 Recombinant Protein

Brand:  R&D Systems™ 501-RL-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465234

  • £884.00 / 50µg


Estimated Shipment
29-06-2018
Add to basket

Description

Description

Source: E. coli-derived, Val117-Ser268, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MES, NaCl, EDTA, DTT and PEG 8000.
Entrez Gene ID: 24494
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in MES, NaCl, EDTA, DTT and PEG 8000 with BSA as a carrier protein.
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Yes
24494
IL-1 beta/IL-1F2
50μg
Unlabeled
Videos