missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IL-1 beta/IL-1F2 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 501-RL-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13445234

  • £352.24 / 10µg

Estimated Shipment
12-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Rat IL-1 beta/IL-1F2 Protein is derived from E. coli. The Recombinant Rat IL-1 beta/IL-1F2 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in MES, NaCl, EDTA, DTT and PEG 8000 with BSA as a carrier protein.
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived rat IL-1 beta/IL-1F2 protein Val117-Ser268,with an N-terminal Met
Unconjugated
24494
IL-1 beta/IL-1F2
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents