missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Rat IL-1 beta/IL-1F2 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 24494 |
Molecular Weight (g/mol) | 17 kDa |
Name | IL-1 beta/IL-1F2 |
Recombinant | Yes |
Description
The Recombinant Rat IL-1 beta/IL-1F2 Protein is derived from E. coli. The Recombinant Rat IL-1 beta/IL-1F2 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
17 kDa | |
Yes |
24494 | |
IL-1 beta/IL-1F2 | |
E. coli-derived rat IL-1 beta/IL-1F2 protein Val117-Ser268,with an N-terminal Met |