missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IFN-gamma Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 585-IF-100/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13435584

  • £273.42 / 100µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Rat IFN-gamma Protein is derived from E. coli. The Recombinant Rat IFN-gamma Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in Tris, NaCl and PEG 3350.
15.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Unlabeled
25712
IFN-gamma
100μg
3 months from date of receipt, 2 to 8° C as supplied.
Yes
Videos
Documents