missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IFN-gamma ELISpot Kit

highly sensitive, microplate-based assays for the detection of cytokine secreting cells

Brand:  R&D Systems™ XEL585

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13443306

  • £2219.94 / Pack of 6

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • Hig
Specifications

Specifications

Diagnostics
Rat IFN-gamma ELISpot Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Six 96-well PVDF-backed plates coated w/pAb for rat IFNG, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT; Positive control
Whole Cells
Six 96-well plates
Videos