missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Rat GM-CSF Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifica
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 116630 |
Molecular Weight (g/mol) | 14.7 kDa |
Name | GM-CSF |
Recombinant | Yes |
Descrizione
The Recombinant Rat GM-CSF Protein is derived from E. coli. The Recombinant Rat GM-CSF Protein has been validated for the following applications: Bioactivity.Specifica
Unconjugated | |
14.7 kDa | |
Yes |
116630 | |
GM-CSF | |
E. coli-derived rat GM-CSF protein Ala1-Lys127,with and without an N-terminal Met |