missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat GFR alpha-1/GDNF R alpha-1 Fc Chimera Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity

Brand:  R&D Systems™ 560-GR-100

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13445514

  • £245.53 / 100µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Rat GFR alpha-1 Fc Chimera Protein is derived from NS0. The Recombinant Rat GFR alpha-1 Fc Chimera Protein has been validated for the following applications: Bioactivity, Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
74 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
NS0
Unlabeled
25454
GFR alpha-1/GDNF R alpha-1 Fc Chimera
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos