missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Rat FGF-BP Recombinant Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 64535 |
Molecular Weight (g/mol) | 26 kDa |
Name | FGF-BP |
Quantity | 25μg |
Description
The Recombinant Rat FGF-BP Protein is derived from NS0. The Recombinant Rat FGF-BP Protein has been validated for the following applications: Binding Activity.Specifications
Unconjugated | |
26 kDa | |
25μg | |
Mouse myeloma cell line,NS0-derived rat FGF-BP protein Glu21-Cys238,with a C-terminal Asp and 10-His tag |
64535 | |
FGF-BP | |
Yes |