missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat Fas Ligand/TNFSF6 Recombinant Protein

Brand:  R&D Systems™ 1614-FL-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13439803

  • £341.89 / 25µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Source: Mouse myeloma cell line, NS0-derived, Leu104-Leu278, with an N-terminal 7-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 25385
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
21 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Yes
25385
Fas Ligand/TNFSF6
25μg
Unlabeled
Videos