missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat CXCL1/GRO alpha/KC/CINC-1 Recombinant Protein

Brand:  R&D Systems™ 515-CN-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465314

  • £153.12 / 10µg


Estimated Shipment
02-01-2019
Add to basket

Description

Description

Source: E. coli-derived, Ala25-Lys96
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 81503
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
7.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Yes
81503
CXCL1/GRO alpha/KC/CINC-1
10μg
Unlabeled
Videos