missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rat CX3CL1/Fractalkine (aa 22-100) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Gene ID (Entrez) | 89808 |
---|---|
Molecular Weight (g/mol) | 9.0 kDa |
Name | CX3CL1/Fractalkine Chemokine Domain, aa 22-100 |
Quantity | 25μg |
Source | E. coli-derived rat CX3CL1/Fractalkine protein Leu22-Gly100 |
Description
The Recombinant Rat CX3CL1/Fractalkine (aa 22-100) Protein is derived from E. coli. The Recombinant Rat CX3CL1/Fractalkine (aa 22-100) Protein has been validated for the following applications: Bioactivity.Specifications
89808 | |
CX3CL1/Fractalkine Chemokine Domain, aa 22-100 | |
E. coli-derived rat CX3CL1/Fractalkine protein Leu22-Gly100 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
9.0 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |