missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Rat CNTF Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 25707 |
Molecular Weight (g/mol) | 22.8 kDa |
Name | CNTF |
Recombinant | Yes |
Description
The Recombinant Rat CNTF Protein is derived from E. coli. The Recombinant Rat CNTF Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
22.8 kDa | |
Yes |
25707 | |
CNTF | |
E. coli-derived rat CNTF protein Ala2-Met200 |