missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat CNTF R alpha Recombinant Protein

Brand:  R&D Systems™ 558-CR-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465504

  • £377.53 / 50µg


Estimated Shipment
29-10-2018
Add to basket

Description

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala19-Pro346 & Gln23-Pro346
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 39.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 313173
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
39.1 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Yes
313173
CNTF R alpha
50μg
Unlabeled
Videos