missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat CD3+ T Cell Enrichment Columns

B cells bind to the anti-Ig coated beads and monocytes bind via Fc interactions for resulting T cell-rich eluate

Brand:  R&D Systems™ RTCC-10

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462636

  • £396.52 / Pack of 10


Estimated Shipment
28-02-2019
Add to basket

Includes: Cell Selection Columns, 10X Column Buffer, Antibody Cocktail Vials

Description

Description

Purifies CD3+ T cells to 85 to 95%
Based upon gravitational flow
Simple procedure
No specialized equipment required
Total CD3+ cell recovery ranges between 30% and 50% of initial CD3+ cells loaded;

Specifications

Specifications

10/Pack
Rat CD3+ T Cell Enrichment Columns
Videos