missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat C-Reactive Protein/CRP Recombinant Protein

Brand:  R&D Systems™ 1744-CR-200/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13428693

  • £267.54 / 200µg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Source: Mouse myeloma cell line, NS0-derived, His20-Ser230, with a C-terminal 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 24 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 25419
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
24 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Unlabeled
25419
C-Reactive Protein/CRP
200μg
Homopentamer
Yes
Videos
Documents