missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat Agrin Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 550-AG-100/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13445474

  • £368.29 / 100µg

Estimated Shipment
21-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Rat Agrin Protein is derived from Sf 21 (baculovirus). The Recombinant Rat Agrin Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
90 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Unlabeled
25592
Agrin
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos