missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Rat Agrin Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 25592 |
Molecular Weight (g/mol) | 90 kDa |
Name | Agrin |
Quantity | 100μg |
Description
The Recombinant Rat Agrin Protein is derived from Sf 21 (baculovirus). The Recombinant Rat Agrin Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
90 kDa | |
100μg | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived rat Agrin protein Ala1153-Pro1959 (Pro1788-Ser1798 del),with an N-terminal Met and 6-His tag |
25592 | |
Agrin | |
Yes |