missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Rabbit Isopeptidase T/USP5 Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Conjugate | Unconjugated |
---|---|
Formulation | X mg/mL (X µM) in 50 mM HEPES pH 8.0, 150 mM NaCl, 1 mM DTT |
Name | Isopeptidase T/USP5 |
Source | Rabbit erythrocyte-derived Isopeptidase T/USP5 protein |
Description
Rabbit Isopeptidase T/USP5 is a Ubiquitin-specific deconjugating enzyme. Reaction conditions will need to be optimized for each specific application. We recommend an initial Rabbit Isopeptidase T/USP5 concentration of 10-100 nM.Specifications
Unconjugated | |
X mg/mL (X µM) in 50 mM HEPES pH 8.0, 150 mM NaCl, 1 mM DTT | |
Isopeptidase T/USP5 | |
Rabbit erythrocyte-derived Isopeptidase T/USP5 protein |