Learn More
R&D Systems™ Rabbit Isopeptidase T/USP5 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ E-320-025
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13434985
  • £241.08 / 25µg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
The Rabbit Isopeptidase T/USP5 Protein is derived from Rabbit Erythrocytes. The Rabbit Isopeptidase T/USP5 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Formulation cannot be determined without the BulkLotPrefix
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Rabbit
Unlabeled
Isopeptidase T/USP5
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.