missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rabbit IL-1 beta/IL-1F2 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 7464-RB-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13436194

  • £265.33 / 10µg

Estimated Shipment
08-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Rabbit IL-1 beta/IL-1F2 Protein is derived from E. coli. The Recombinant Rabbit IL-1 beta/IL-1F2 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in MES, NaCl, DTT and EDTA with BSA as a carrier protein.
IL-1 beta/IL-1F2
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
17.5 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rabbit
<0.1 EU per 1μg of the protein by the LAL method.
Yes
Videos