missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Rabbit IL-1 beta/IL-1F2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Molecular Weight (g/mol) | 17.5 kDa |
---|---|
Name | IL-1 beta/IL-1F2 |
Quantity | 10μg |
Source | E. coli-derived rabbit IL-1 beta/IL-1F2 protein Ala117-Ser268 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Description
The Recombinant Rabbit IL-1 beta/IL-1F2 Protein is derived from E. coli. The Recombinant Rabbit IL-1 beta/IL-1F2 Protein has been validated for the following applications: Bioactivity.Specifications
17.5 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
Recombinant |
IL-1 beta/IL-1F2 | |
E. coli-derived rabbit IL-1 beta/IL-1F2 protein Ala117-Ser268 | |
<0.1 EU per 1μg of the protein by the LAL method. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |