missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Proteome Purify™ 2 Human Serum Protein Immunodepletion Resin

A single-use resin that depletes serum or plasma of albumin and IgG to improve visualization of target proteins

Brand:  R&D Systems™ IDR002-040

Code : Z1

Additional Details : Weight : 0.00040kg

 View more versions of this product

Product Code. 13427955

  • £940.80 / 40mL


Estimated Shipment date
from Supplier 31-03-2020
Add to basket

Includes: Immunodepletion Resin, Spin-X Filter Units

Description

Description

Allows user to increase protein load or concentration
Removal allows visualization of proteins that co-migrate or co-purify with the high-abundance proteins
Kit removes proteins representing approximately 60% of the protein contained in serum and plasma

Specifications

Specifications

40 Tests
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Proteome Purify 2 Human Serum Protein Immunodepletion Resin
Videos
Documents