missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Proteome Purify™ 12 Human Serum Protein Immunodepletion Resin

A single-use alternative to existing column or spin cartridge systems to deplete serum or plasma of high abundance proteins

Brand:  R&D Systems™ IDR012-040

Code : Z1

Additional Details : Weight : 0.00040kg

 View more versions of this product

Product Code. 13447955

  • £921.69 / 40mL

Estimated Shipment date
from Supplier 17-12-2019

Please to purchase this item. Need a web account? Register with us today!


Includes: Immunodepletion Resin, Spin-X Filter Units

Description

Description

Depletes samples of the 12 high-abundance proteins comprising approximately 95% of proteins in human serum or plasma
Depletes samples of albumin, IgA, IgG and IgM, Fibrinogen, Transferrin, Haptoglobin and others
Removal allows visualization of proteins

Specifications

Specifications

40 Tests
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Proteome Purify 12 Human Serum Protein Immunodepletion Resin
Videos
Documents