missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Porcine TGF-beta 1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 397078 |
Name | TGF-beta 1 |
Source | Porcine |
Description
The Porcine TGF-beta 1 Protein is derived from Porcine Platelets. The Porcine TGF-beta 1 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
397078 | |
TGF-beta 1 | |
Porcine |