missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Porcine TGF-beta 1 Protein

Brand:  R&D Systems™ 101-B1-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458313

  • £1602.46 / 10µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Source: Porcine platelet-derived
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 397078
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
TGF-beta 1
10μg
Disulfide-linked homodimer
397078
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Porcine
Unlabeled
Videos