Learn More
R&D Systems™ Porcine TGF-beta 1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 101-B1-010
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448313
  • £1884.54 / 10µg
Call for latest availability
+44 (0)1509 555 500
Description
Description
The Porcine TGF-beta 1 Protein is derived from Porcine Platelets. The Porcine TGF-beta 1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
TGF-beta 1
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
397078
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Porcine
Disulfide-linked homodimer
Unlabeled