missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Porcine TGF-beta 1 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 101-B1-001/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438313

  • £287.77 / 1µg

Estimated Shipment date
from Supplier 27-09-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Porcine TGF-beta 1 Protein is derived from Porcine Platelets. The Porcine TGF-beta 1 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
TGF-beta 1
1μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
397078
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Porcine
Disulfide-linked homodimer
Videos
Documents