missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Porcine IL-5 Recombinant Protein

Brand:  R&D Systems™ 3137-PL-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462504

  • £320.00 / 10µg


Estimated Shipment
29-06-2018
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ile20-Ser134, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 397409
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
14 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Porcine
Unlabeled
397409
IL-5
10μg
Disulfide-linked homodimer
Yes
Videos